Anti-FOXP1 Rabbit Polyclonal Antibody

Supplier: Abcam
AB227788-1ML AB227788-100UL AB227788-500UL
ABCA_AB227788-1MLEA 1003 USD
ABCA_AB227788-1ML ABCA_AB227788-100U ABCA_AB227788-500U
Anti-FOXP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to FOXP1 - C-terminal.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: FOXP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR