Anti-TAGLN/Transgelin Human Monoclonal Antibody [clone: EPR21206]

Supplier: Abcam
AB213273-1ML AB213273-100UL
ABCA_AB213273-1MLEA 6041 USD
ABCA_AB213273-1ML ABCA_AB213273-100U
Anti-TAGLN/Transgelin Human Monoclonal Antibody [clone: EPR21206]
Antibodies
Human monoclonal [EPR21206] to TAGLN/Transgelin.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: TAGLN/Transgelin
Clonality: Monoclonal
Clone: EPR21206
Conjugation:
Epitope:
Host: Human
Isotype: IgG1
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR