Anti-IgE Mouse Monoclonal Antibody [clone: 15.3]

Supplier: Abcam
AB37425-25UG AB37425-250UG
ABCA_AB37425-25UGEA 257 USD
ABCA_AB37425-25UG ABCA_AB37425-250UG
Anti-IgE Mouse Monoclonal Antibody [clone: 15.3]
Antibodies
Mouse IgE monoclonal [15.3] - Isotype Control.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Control
Antigen: IgE
Clonality: Monoclonal
Clone: 15.3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgE
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR