Anti-GC Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A74043-100UL A74043-50UL
77848-957EA 330 USD
77848-957 77848-958
Anti-GC Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA:1:2000 to 1:5000, WB:1:500 to 1:2000, IHC:1:50 to 1:200

Type:
Antigen: GC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR