Anti-CENPE Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A74026-100UL A74026-50UL
77848-930EA 330 USD
77848-930 77848-931
Anti-CENPE Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA:1:1000 to 1:5000, IHC:1:50 to 1:200

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR