Anti-EMD Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A74544-100UL A74544-50UL
77849-795EA 330 USD
77849-795 77849-796
Anti-EMD Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:10000, WB: 1:1000 to 1:5000

Type:
Antigen: EMD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR