Anti-SLC25A2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A74042-100UL A74042-50UL
77848-955EA 330 USD
77848-955 77848-956
Anti-SLC25A2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:20 to 1:100

Type:
Antigen: SLC25A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR