Anti-MC4R Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A28283-100UG A28283-50UG
77742-864EA 330 USD
77742-864 77859-558
Anti-MC4R Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: MC4R
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR