Anti-H2AFZ Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A24298-100UL A24298-50UL
77739-374EA 350 USD
77739-374 77856-694
Anti-H2AFZ Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IF: 1:50 to 1:200

Type:
Antigen: H2AFZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR