Anti-YWHAZ Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A28637-100UG A28637-50UG
77743-197EA 330 USD
77743-197 77859-813
Anti-YWHAZ Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: YWHAZ
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR