Anti-IGFBP6 Rabbit Recombinant polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: AFG Bioscience
A25980-100UG A25980-50UG
77740-761EA 330 USD
77740-761 77857-937
Anti-IGFBP6 Rabbit Recombinant polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
0

Type:
Antigen: IGFBP6
Clonality: Recombinant polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR