Anti-GC Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A42738-100UG A42738-50UG
77812-018EA 330 USD
77812-018 77812-019
Anti-GC Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: GC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR