Anti-VAMP5 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A38225-100UG A38225-50UG
77804-235EA 330 USD
77804-235 77804-236
Anti-VAMP5 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: VAMP5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR