Anti-CLIC1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A51123-100UL A51123-50UL
77816-580EA 330 USD
77816-580 77816-581
Anti-CLIC1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:50 to 1:200

Type:
Antigen: CLIC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR