Anti-FEZ1 Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A39788-100UG A39788-50UG
77806-707EA 330 USD
77806-707 77806-708
Anti-FEZ1 Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: FEZ1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR