Anti-RNF213 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A57405-100UG A57405-50UG
77828-063EA 330 USD
77828-063 77828-064
Anti-RNF213 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: RNF213
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR