Anti-MYC Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A38340-100UG A38340-50UG
77804-415EA 330 USD
77804-415 77804-416
Anti-MYC Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: MYC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR