Anti-TET3 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A36396-100UG A36396-50UG
77802-245EA 330 USD
77802-245 77802-246
Anti-TET3 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: TET3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR