Anti-HSPA8 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A39725-100UG A39725-50UG
77806-592EA 330 USD
77806-592 77806-593
Anti-HSPA8 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: HSPA8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR