Anti-SMCP Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A35199-100UG A35199-50UG
77800-331EA 330 USD
77800-331 77800-332
Anti-SMCP Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: SMCP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR