Anti-VAMP7 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A38232-100UG A38232-50UG
77804-248EA 330 USD
77804-248 77804-249
Anti-VAMP7 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: VAMP7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR