Anti-TEAD1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A36362-100UL A36362-50UL
77802-191EA 330 USD
77802-191 77802-192
Anti-TEAD1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:500, IF: 1:50 to 1:200

Type:
Antigen: TEAD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR