Anti-SMAD3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A39767-50UL A39767-100UL
77806-670EA 225 USD
77806-670 77806-669
Anti-SMAD3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:25 to 1:100

Type:
Antigen: SMAD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR