Anti-SMN1 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A54561-100UG A54561-50UG
77822-731EA 330 USD
77822-731 77822-732
Anti-SMN1 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: SMN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR