Anti-afp Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A50618-100UG A50618-50UG
77815-636EA 330 USD
77815-636 77815-637
Anti-afp Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: afp
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Aspergillus giganteus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR