Anti-STK33 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A66632-100UG A66632-50UG
77835-328EA 330 USD
77835-328 77835-329
Anti-STK33 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500 to 1:2000, IHC:1:20 to 1:200, IF:1:50 to 1:200

Type:
Antigen: STK33
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR