Anti-TMEM150A Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A59842-100UG A59842-50UG
77832-656EA 330 USD
77832-656 77832-657
Anti-TMEM150A Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IF: 1:50 to 1:200

Type:
Antigen: TMEM150A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR