Tetraethylammonium hydroxide 35% in water

Supplier: TCI America
Danger

Synonyms: Tetrylammonium hydroxide

T2589-25ML T2589-500ML
TCT2589-25MLEA 29 USD
TCT2589-25ML TCT2589-500ML
Tetraethylammonium hydroxide 35% in water
Tetraethylammonium hydroxide

CAS Number: 77-98-5
MDL Number: MFCD00009024
Molecular Formula: C8H21NO
Molecular Weight: 147.26
Form: Clear Liquid
Color: Colorless

Formula: C₈H₂₁NO
MW: 147.26 g/mol
MDL Number: MFCD00009024
CAS Number: 77-98-5
EINECS: 201-073-3
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e725f8d157a5f2715c1c65bae9a4ae45454a527&vid=850fe737379d16875e2a1b521d674ffe0bade77c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR