Tetramethylammonium hydroxide ∼25% in water

Supplier: TCI America
Danger

T1460-500ML T1460-25ML
TCT1460-500MLEA 87.31 USD
TCT1460-500ML TCT1460-25ML
Tetramethylammonium hydroxide ∼25% in water
Tetramethylammonium hydroxide

CAS Number: 75-59-2
MDL Number: MFCD00008280
Molecular Formula: C4H13NO
Molecular Weight: 91.15
Form: Clear Liquid
Color: Colorless


Other Product Names / Synonyms: TMAH

Formula: C₄H₁₃NO
MW: 91.15
Density: 1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008280
CAS Number: 75-59-2
EINECS: 200-882-9
UN: 1835
ADR: 8 II
Merck Index: 14,09224
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6adfddc35a1152ba6a61b2b334241c695a2980f7&vid=112fb51b484f163b661cd0edde34af7f41c1dcf8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR