Thymine ≥98.0%
Supplier: TCI America
Synonyms:
5-Methylpyrimidine-2,4(1H,3H)-dione, 5-Methyluracil
T0234-25G
T0234-5G
T0234-100G
TCT0234-025GEA
36
USD
TCT0234-025G
TCT0234-5G
TCT0234-100G
Thymine ≥98.0%
Thymine
CAS Number: 65-71-4
MDL Number: MFCD00006026
Molecular Formula: C5H6N2O2
Molecular Weight: 126.12
Purity/Analysis Method:
Product Categories: Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: 5-Methyluracil
Formula:
C₅H₆N₂O₂ MW: 126.11 g/mol Boiling Pt: 389 Melting Pt: 218 Density: 1.454 g/cm³ (25 °C) |
MDL Number:
MFCD00006026 CAS Number: 65-71-4 EINECS: 200-616-1 Merck Index: 12,09539 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...