Theobromine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 3,7-Dimethyl-3,7-dihydro-1H-purine-2,6-dione, 3,7-Dimethylxanthine, 3,7-Dihydro-3,7-dimethyl-1H-purine-2,6-dione, 2,6-Dihydroxy-3,7-dimethylpurine

T0178-25G T0178-500G T0178-100G
TCT0178-025GEA 61.96 USD
TCT0178-025G TCT0178-500G TCT0178-100G
Theobromine ≥98.0% (by HPLC, titration analysis)
Theobromine

CAS Number: 83-67-0 MDL Number: MFCD00022830 Molecular Formula: C7H8N4O2 Molecular Weight: 180.17 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Color: White Melting point (°C): 350


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Crude Medicine Ingredients for Research and Experimental Use, Other Pharmaceutical Ingredients, Alkaloids (Phytochemicals), Purine Alkaloids Other Product Names / Synonyms: 3,7-Dimethylxanthine

Formula: C₇H₈N₄O₂
MW: 180.17 g/mol
Melting Pt: 350 °C
Density: 1.419 g/cm³ (25 °C)
MDL Number: MFCD00022830
CAS Number: 83-67-0
EINECS: 201-494-2
Merck Index: 12,09418
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eff53f59620a1db183f5f7fa2b6d64fe205c5c9f&vid=eff53f59620a1db183f5f7fa2b6d64fe205c5c9f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR