Tetramethylthiuram monosulfide ≥98.0%

Supplier: TCI America
Warning

Synonyms: Bis(dimethylthiocarbamyl) sulphide, Bis(dimethylthiocarbamyl) sulfide

T0157-25G T0157-500G
TCT0157-025GEA 31.52 USD
TCT0157-025G TCT0157-500G
Tetramethylthiuram monosulfide ≥98.0%
Tetramethylthiuram monosulphide

CAS Number: 97-74-5 MDL Number: MFCD00014870 Molecular Formula: C6H12N2S3 Molecular Weight: 208.36 Purity/Analysis Method: 98.0% (GC) Form: Crystal Color: Pale Yellow Melting point (°C): 110 Flash Point (°C): 156

Formula: C₆H₁₂N₂S₃
Melting Pt: 110 °C
Density: 1.37 g/cm³ (20 °C)
Flash Pt: 156 °C
MDL Number: MFCD00014870
CAS Number: 97-74-5
EINECS: 202-605-7
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed63a98b8d44c380a166c098b952d4180896a1df&vid=ed63a98b8d44c380a166c098b952d4180896a1df&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR