Quinoline ≥97.0%

Supplier: TCI America
Danger

Q0085-25G Q0085-500G
TCQ0085-25GEA 21.26 USD
TCQ0085-25G TCQ0085-500G
Quinoline ≥97.0%
Quinoline

CAS Number: 91-22-5 MDL Number: MFCD00006736 Molecular Formula: C9H7N Molecular Weight: 129.16 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 238 Melting point (°C): -15 Flash Point (°C): 106 Specific Gravity (20/20): 1.09

Formula: C₉H₇N
MW: 129.16 g/mol
Boiling Pt: 238 °C
Melting Pt: –15 °C
Density: 1.09 (20 °C)
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00006736
CAS Number: 91-22-5
EINECS: 202-051-6
UN: 2656
ADR: 6.1,III
Merck Index: 13,08160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=158670e9b21d838d9ea9c4f43510f7ba095f5f2d&vid=cbc3fd17586f088699d1dc82ee081d62c07bb462&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR