(-)-Quinine ≥98.0%
Supplier: TCI America
Synonyms:
(R)-[(2S,4S,5R)-1-Aza-5-vinylbicyclo[2.2.2]oct-2-yl](6-methoxyquinolin-4-yl)methanol, Quinine, (R)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol
Q0028-25G
Q0028-100G
TCQ0028-025GEA
106.8
USD
TCQ0028-025G
TCQ0028-100G
(-)-Quinine ≥98.0%
(-)-Quinine
CAS Number: 130-95-0
MDL Number: MFCD00198096
Molecular Formula: C20H24N2O2
Molecular Weight: 324.42
Purity/Analysis Method:
Formula:
C₂₀H₂₄N₂O₂ Melting Pt: 177 °C |
MDL Number:
MFCD00198096 CAS Number: 130-95-0 EINECS: 205-003-2 Merck Index: 12,08245 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...