(R)-(-)-Hydratropic acid ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
(R)-(-)-2-Phenylpropanoic acid, (R)-(-)-2-Phenylpropionic acid, (R)-2-Phenylpropanoic acid
CAS Number: 7782-26-5
MDL Number: MFCD00063140
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (R)-(-)-Hydratropic Acid
Formula:
C₉H₁₀O₂ MW: 150.18 g/mol Boiling Pt: 257 Melting Pt: 29…32 °C Density: 1.1 g/cm³ (20 °C) Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00063140 CAS Number: 7782-26-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...