Dibutyl phthalate ≥97.0%

Supplier: TCI America
Danger

Synonyms: DBP, Phthalic acid dibutyl ester

P0292-500ML P0292-25ML
TCP0292-500MLEA 33.73 USD
TCP0292-500ML TCP0292-25ML
Dibutyl phthalate ≥97.0%
Dibutyl phthalate

CAS Number: 84-74-2 MDL Number: MFCD00009441 Molecular Formula: C16H22O4 Molecular Weight: 278.35 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Boiling point (°C): 340 Melting point (°C): -35 Flash Point (°C): 157 Specific Gravity (20/20): 1.05


Product Categories: Materials Chemistry, Resin Additives, Analytical Science, Phthalates (Environmental Endocrine Disruptors) Other Product Names / Synonyms: Phthalic Acid Dibutyl Ester, DBP

Formula: C₁₆H₂₂O₄
MW: 278.35 g/mol
Boiling Pt: 340 °C
Melting Pt: –35 °C
Density: 1.05 (20 °C)
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00009441
CAS Number: 84-74-2
EINECS: 201-557-4
UN: 3082
ADR: 9,III
REACH: 01-2119493042-44
Merck Index: 13,01586
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db74ff1cae12482049d90843967d2a8c47b6da42&vid=db74ff1cae12482049d90843967d2a8c47b6da42&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR