Phenyl dichlorophosphate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Phenyl phosphorodichloridate
P0209-500G
P0209-25G
TCP0209-500GEA
316.71
USD
TCP0209-500G
TCP0209-25G
Phenyl dichlorophosphate ≥98.0% (by GC, titration analysis)
Phenyl dichlorophosphate
CAS Number: 770-12-7
MDL Number: MFCD00002067
Molecular Formula: C6H5Cl2O2P
Molecular Weight: 210.98
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Phosphorylation, Nucleic Acid Synthesis
Other Product Names / Synonyms: Phenylphosphoric Acid Dichloride, Phenyl Phosphorodichloridate, Phenylphosphoryl Dichloride
Formula:
C₆H₅Cl₂O₂P MW: 210.98 g/mol Boiling Pt: 240 °C Density: 1.42 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00002067 CAS Number: 770-12-7 EINECS: 212-220-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...