O-Phenylenediamine ≥98.0%

Supplier: TCI America
Danger

Synonyms: 1,2-Phenylendiamine, 1,2-Phenylenediamine, 1,2-Diaminobenzene, OPD

P0168-500G P0168-25G P0168-100G
TCP0168-500GEA 133 USD
TCP0168-500G TCP0168-25G TCP0168-100G
O-Phenylenediamine ≥98.0%
o-Phenylenediamine

CAS Number: 95-54-5 MDL Number: MFCD00007721 Molecular Formula: C6H8N2 Molecular Weight: 108.14 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Boiling point (°C): 265 Melting point (°C): 103 Flash Point (°C): 156


Product Categories: Synthetic Organic Chemistry, Amines (Multi-component Reaction), Materials Chemistry, Ligands for Functional Metal Complexes Other Product Names / Synonyms: 1,2-Diaminobenzene

Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 257
Melting Pt: 103
Density: 1.21 g/cm³ (20 °C)
Flash Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD00007721
CAS Number: 95-54-5
EINECS: 202-430-6
UN: 1673
ADR: 6.1 III
Merck Index: 12,07438
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eec6a0126029e74a3a232b4fbde4bd14a20588f1&vid=eec6a0126029e74a3a232b4fbde4bd14a20588f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR