n-Pentacosane ≥97.0%

Supplier: TCI America

Synonyms: N-pentacosane

P0139-5G P0139-1G
TCP0139-005GEA 214.77 USD
TCP0139-005G TCP0139-1G
n-Pentacosane ≥97.0%
n-Pentacosane

CAS Number: 629-99-2 MDL Number: MFCD00009353 Molecular Formula: C25H52 Molecular Weight: 352.69 Purity/Analysis Method: 97.0% (GC) Form: Crystal Boiling point (°C): 254 Melting point (°C): 54 Flash Point (°C): 120

Formula: H₃C(CH₂)₂₃CH₃
MW: 352.69 g/mol
Boiling Pt: 254 °C
Melting Pt: 54 °C
Density: 0.8012 g/cm³ (20 °C)
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00009353
CAS Number: 629-99-2
EINECS: 211-123-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4542ffa9f67fbc31b434718cc7684354ca1cbf46&vid=ef83affac2762ebbd2f745f812d4a41dccb1408e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR