Sudan IV

Supplier: TCI America
Warning

Synonyms: Oil Red, 1-((2-Methyl-4-(o-tolyldiazenyl)phenyl)diazenyl)naphthalen-2-ol, Scarlet Red, Oil Red BB, 1-({2-Methyl-4-[(2-methylphenyl)diazenyl]-phenyl}diazenyl)-2-naphthol, Scarlet R, Biebrich scarlet R, Lipid Crimson, Scarlet Red Scharlach, Oil Red IV, Fat Red B, Solvent Red 24, 1-(2-Methyl-4-(2-methylphenylazo)phenylazo)-2-naphthol

O0100-25G O0100-100G
TCO0100-025GEA 34.96 USD
TCO0100-025G TCO0100-100G
Sudan IV
Sudan IV

CAS Number: 85-83-6 MDL Number: MFCD00003893 Molecular Formula: C24H20N4O Molecular Weight: 380.45 Form: Crystal Color: Yellow Red


Other Product Names / Synonyms: Bieblich Scarlet R, Oil Red, Scarlet Red, Scharlach Red, Solvent Red 24

Formula: C₂₄H₂₀N₄O
MW: 380.45 g/mol
Melting Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00003893
CAS Number: 85-83-6
EINECS: 201-635-8
Merck Index: 12,08538
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=208ba432e564eb0b71084ff79b1e9d2229454f3e&vid=208ba432e564eb0b71084ff79b1e9d2229454f3e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR