2-Nitrotoluene ≥99.0%
Supplier: TCI America
|
Danger
|
N0275-500G
N0275-25G
TCN0275-500GEA
75
USD
TCN0275-500G
TCN0275-25G
2-Nitrotoluene ≥99.0%
2-Nitrotoluene
CAS Number: 88-72-2
MDL Number: MFCD00007157
Molecular Formula: C7H7NO2
Molecular Weight: 137.14
Purity/Analysis Method:
Formula:
C₇H₇NO₂ MW: 137.14 g/mol Boiling Pt: 221.7 Melting Pt: -1 Density: 1.17 (20 °C) Flash Pt: 106 Storage Temperature: Ambient |
MDL Number:
MFCD00007157 CAS Number: 88-72-2 EINECS: 201-853-3 UN: 1664 ADR: 6.1 II Merck Index: 12,06748 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...