5-Nitro-o-toluidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

N0115-500G N0115-25G
TCN0115-500GEA 607 USD
TCN0115-500G TCN0115-25G
5-Nitro-o-toluidine ≥98.0% (by GC, titration analysis)
5-Nitro-o-toluidine

CAS Number: 99-55-8 MDL Number: MFCD00007741 Molecular Formula: C7H8N2O2 Molecular Weight: 152.15 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Melting point (°C): 109


Other Product Names / Synonyms: 2-Amino-4-nitrotoluene, 5-Nitro-o-toluidine

Formula: C₇H₈N₂O₂
Melting Pt: 109 °C
Density: 1.365 g/cm³ (15 °C)
MDL Number: MFCD00007741
CAS Number: 99-55-8
EINECS: 202-765-8
UN: 2660
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a7ceda20bf2d975bc4bcb72d4897b84c33bfa87&vid=4a7ceda20bf2d975bc4bcb72d4897b84c33bfa87&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR