4-Methylcyclohexanone ≥98.0%
Supplier: TCI America
M0199-500ML
M0199-25ML
TCM0199-500MLEA
294
USD
TCM0199-500ML
TCM0199-25ML
4-Methylcyclohexanone ≥98.0%
4-Methylcyclohexanone
CAS Number: 589-92-4
MDL Number: MFCD00001643
Molecular Formula: C7H12O
Molecular Weight: 112.17
Purity/Analysis Method:
Formula:
C₇H₁₂O Boiling Pt: 171 °C Melting Pt: –41 °C Density: 0.92 (20 °C) Flash Pt: 48 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001643 CAS Number: 589-92-4 EINECS: 209-665-3 UN: 2297 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...