Methylcyclohexane ≥99.0%
Supplier: TCI America
|
Danger
|
M0190-500ML
M0190-25ML
TCM0190-500MLEA
58.69
USD
TCM0190-500ML
TCM0190-25ML
Methylcyclohexane ≥99.0%
Methylcyclohexane
CAS Number: 108-87-2
MDL Number: MFCD00001497
Molecular Formula: C7H14
Molecular Weight: 98.19
Purity/Analysis Method:
Product Categories: Materials Chemistry, Fuel Cell Research Reagents
Other Product Names / Synonyms: Hexahydrotoluene, MCH
Formula:
C₆H₁₁CH₃ MW: 98.19 g/mol Boiling Pt: 100 °C Melting Pt: –127 °C Density: 0.77 (20 °C) Flash Pt: –4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001497 CAS Number: 108-87-2 EINECS: 203-624-3 UN: 2296 ADR: 3,II Merck Index: 13,06074 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...