2-Mercaptoethanol ≥98.0%

Supplier: TCI America
Danger

Synonyms: beta-Mercaptoethanol, 2-Hydroxyethyl mercaptan, β-Mercaptoethanol

M0058-500G M0058-25G
TCM0058-500GEA 77.05 USD
TCM0058-500G TCM0058-25G
2-Mercaptoethanol ≥98.0%
2-Mercaptoethanol

CAS Number: 60-24-2 MDL Number: MFCD00004890 Molecular Formula: C2H6OS Molecular Weight: 78.13 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 158 Melting point (°C): -100 Flash Point (°C): 74 Specific Gravity (20/20): 1.12


Product Categories: Bioscience, Antioxidants, Polyamine Synthesis Reagents, Functionality Creating Reagents, Disulfide Reducing Agents Other Product Names / Synonyms: 2-Hydroxyethyl Mercaptan, Thioglycol

Formula: HSCH₂CH₂OH
MW: 78.14 g/mol
Boiling Pt: 158 °C
Melting Pt: –100 °C
Density: 1.12 (20 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00004890
CAS Number: 60-24-2
EINECS: 200-464-6
UN: 2966
ADR: 6.1,II
Merck Index: 13,05894
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b2de4a27b611bfba008542b0f8d01b41c1f5970&vid=4b2de4a27b611bfba008542b0f8d01b41c1f5970&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR