(R)-(+)-Limonene ≥99.0%
Supplier: TCI America
|
Danger
|
CAS Number: 5989-27-5
MDL Number: MFCD00062991
Molecular Formula: C10H16
Molecular Weight: 136.24
Purity/Analysis Method:
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Terpenoids (Phytochemicals), Monocyclic Monoterpenes
Other Product Names / Synonyms: (+)-p-Mentha-1,8-diene
Formula:
C₁₀H₁₆ Boiling Pt: 176 °C Melting Pt: –75 °C Density: 0.84 (20 °C) Flash Pt: 51 °C Storage Temperature: Ambient |
MDL Number:
MFCD00062991 CAS Number: 5989-27-5 EINECS: 227-813-5 UN: 2052 ADR: 3,III Merck Index: 13,05515 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...