Isobutylene oxide (2,2-dimethyloxirane, 1,2-epoxy-2-methylpropane) ≥98.0%

Supplier: TCI America

Synonyms: Isobutylene oxide

I0323-500ML I0323-25ML
TCI0323-500MLEA 796 USD
TCI0323-500ML TCI0323-25ML
Isobutylene oxide (2,2-dimethyloxirane, 1,2-epoxy-2-methylpropane) ≥98.0%
Isobutylene oxide (2,2-Dimethyloxirane, 1,2-Epoxy-2-methylpropane)

CAS Number: 558-30-5 MDL Number: MFCD00066354 Molecular Formula: C4H8O Molecular Weight: 72.11 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 52 Flash Point (°C): -32 Specific Gravity (20/20): 0.80


Other Product Names / Synonyms: 1,2-Epoxyisobutane, 2-Methylpropylene Oxide

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 52 °C
Density: 0.80 (20 °C)
Flash Pt: –32 °C
MDL Number: MFCD00066354
CAS Number: 558-30-5
EINECS: 209-193-8
UN: 2924
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd03e1dbc2536a669a2eb1c80a116073d3c29f42&vid=fd03e1dbc2536a669a2eb1c80a116073d3c29f42&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR