5-Iodo-2'-deoxyuridine (Idoxuridine) ≥99.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidine-2,4(1H,3H)-dione, Idoxuridine, (+)-5-Iodo-2'-deoxyuridine
CAS Number: 54-42-2
MDL Number: MFCD00134656
Molecular Formula: C9H11IN2O5
Molecular Weight: 354.10
Purity/Analysis Method:
Product Categories: Bioscience, Antiviral Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs
Other Product Names / Synonyms: Idoxuridine, 5-Iododesoxyuridine, IDU
Formula:
C₉H₁₁IN₂O₅ MW: 354.10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00134656 CAS Number: 54-42-2 EINECS: 200-207-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...