Isophthalonitrile ≥98.0%

Supplier: TCI America

Synonyms: 1,3-Benzenedicarbonitrile, 1,3-Dicyanobenzene, Benzene-1,3-dicarbonitrile

I0158-25G I0158-250G
TCI0158-025GEA 40 USD
TCI0158-025G TCI0158-250G
Isophthalonitrile ≥98.0%
Isophthalonitrile

CAS Number: 626-17-5 MDL Number: MFCD00001795 Molecular Formula: C8H4N2 Molecular Weight: 128.13 Purity/Analysis Method: 98.0% (GC) Form: Crystal Melting point (°C): 162


Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction) Other Product Names / Synonyms: 1,3-Dicyanobenzene, Isophthalodinitrile

Formula: C₈H₄N₂
Boiling Pt: 288 °C (1013 hPa)
Melting Pt: 162 °C
Density: 4.42 g/cm³ (20 °C)
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00001795
CAS Number: 626-17-5
EINECS: 210-933-7
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b048936235979ba983d356434d967522fd59faa8&vid=b048936235979ba983d356434d967522fd59faa8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR