Isoniazid ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
4-Picolinic acid hydrazide, 4-Pyridinecarboxylic acid hydrazide, Isonicotinic hydrazide, Pyridine-4-carboxylic acid hydrazide, Isonicotinic acid hydrazide
I0138-500G
I0138-25G
I0138-100G
TCI0138-500GEA
180
USD
TCI0138-500G
TCI0138-025G
TCI0138-100G
Isoniazid ≥98.0%
Isoniazid
CAS Number: 54-85-3
MDL Number: MFCD00006426
Molecular Formula: C6H7N3O
Molecular Weight: 137.14
Purity/Analysis Method:
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: Isoniazid
Formula:
C₆H₇N₃O MW: 137.14 g/mol Melting Pt: 172 °C Density: 1.42 g/cm³ (20 °C) Flash Pt: >250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006426 CAS Number: 54-85-3 EINECS: 200-214-6 Merck Index: 13,05205 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...